This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other ZNF80 SIRNA Assays ›
Gene Symbol:
ZNF80
Gene Name:
zinc finger protein 80
Gene Aliases:
pT17
Chromosome Location:
Chr.3: 114211035 - 114237578 on Build GRCh38
UniGene ID:
Hs.271079
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID 3596

Purification
Size
Availability Inventoried
Catalog # AM16708
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: ZNF80
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_007136.3 NP_009067.2 1 1753
XM_017007132.1 XP_016862621.1 1 1441
XM_017007133.1 XP_016862622.1 1 1441
XM_017007134.1 XP_016862623.1 1 1441
XM_017007135.1 XP_016862624.1 1 1441
XM_017007136.1 XP_016862625.1 1 1441
GenBank mRNA BC129925.1 - 1 1278 40121991
X65233.1 - 1 1762

Back To Top

More Information


Panther Classification:

Molecular Function -

C2H2 zinc finger transcription factor

Gene Ontology Categories:

Function(s) Process(es)

transcription, DNA-templated
regulation of transcription, DNA-templated
DNA binding
metal ion binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-blue-67f85b6989-mjsjv:80/100.66.76.16:80.
git-commit: a6f50562c839d11f1e5940093fdcbf4cc66353c1
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.1-Offline