siRNA Details
| Gene Symbol: TSPAN18 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_130783.4 | NP_570139.3 | 6 | 604 | ||
| XM_005253217.3 | XP_005253274.1 | 8 | 788 | |||
| XM_006718372.3 | XP_006718435.1 | 8 | 875 | |||
| XM_006718373.3 | XP_006718436.1 | 7 | 861 | |||
| XM_011520459.2 | XP_011518761.1 | 7 | 751 | |||
| XM_017018530.1 | XP_016874019.1 | 7 | 626 | |||
| XM_017018531.1 | XP_016874020.1 | 6 | 548 | |||
| GenBank mRNA | AK027715.1 | - | 5 | 413 | ||
| AK075547.1 | - | 4 | 337 | |||
| AY358087.1 | - | 5 | 527 | |||
| BC019342.2 | - | 4 | 376 | 3357734 | ||

TSPAN18
Genomic Map