This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other TPTE2 SIRNA Assays ›
Gene Symbol:
TPTE2
Gene Name:
transmembrane phosphoinositide 3-phosphatase and tensin homolog 2
Gene Aliases:
TPIP
Chromosome Location:
Chr.13: 19422877 - 19561574 on Build GRCh38
UniGene ID:
Hs.377488
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID 36400

Purification
Size
Availability Inventoried
Catalog # AM16708
Price 4,126.00
Your Price
Online offer:
4126.0
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: TPTE2
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001141968.1 NP_001135440.1 18, 19 1544
NM_001271850.1 NP_001258779.1 6, 7 823
NM_130785.3 NP_570141.3 17, 18 1454
NM_199254.2 NP_954863.2 20, 21 1685
NR_073485.1 23, 24 1924
NR_073486.1 15, 16 1158
NR_073487.1 14, 15 1222
GenBank mRNA AJ421032.1 - 17, 18 1454
AJ421033.1 - 14, 15 1120
AK301741.1 - 23, 24 1929
AY219890.1 - 20, 21 1685
BC128146.1 - 18, 19 1526 40112547
BC128147.1 - 14, 15 1204
BG207812.1 - 1, 2 76
FJ969729.1 - 6, 7 826

Back To Top

More Information


Panther Classification:

Molecular Function -

protein phosphatase

Gene Ontology Categories:

Function(s) Process(es)

phosphatidylinositol biosynthetic process
peptidyl-tyrosine dephosphorylation
protein tyrosine phosphatase activity
protein tyrosine/serine/threonine phosphatase activity
phosphatidylinositol-3,4,5-trisphosphate 3-phosphatase activity
phosphatidylinositol-3,4-bisphosphate 3-phosphatase activity

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-blue-67f85b6989-wtj7v:80/100.66.79.197:80.
git-commit: a6f50562c839d11f1e5940093fdcbf4cc66353c1
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.1-Offline