siRNA Details
| Gene Symbol: C19orf52 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_138358.2 | NP_612367.1 | 2 | 800 | ||
| GenBank mRNA | BC005075.1 | - | 2 | 777 | 3534875 | |
| BC007244.1 | - | 2 | 790 | 2966591 | ||
| BC011833.2 | - | 2 | 800 | 3941883 | ||

C19orf52
Genomic Map