This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other LRSAM1 SIRNA Assays ›
Gene Symbol:
LRSAM1
Gene Name:
leucine rich repeat and sterile alpha motif containing 1
Gene Aliases:
CMT2P, RIFLE, TAL
Chromosome Location:
Chr.9: 127451486 - 127503501 on Build GRCh38
UniGene ID:
Hs.495188
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID 36678

Purification
Size
Availability Inventoried
Catalog # AM16708
Price 283.00
Your Price
Online offer: 209.42
209.42
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: LRSAM1
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001005373.3 NP_001005373.1 22 2012
NM_001005374.3 NP_001005374.1 21 1717
NM_001190723.2 NP_001177652.1 21 1931
NM_138361.5 NP_612370.3 21 2271
XM_017015283.1 XP_016870772.1 22 3011
XM_017015284.1 XP_016870773.1 22 2007
GenBank mRNA AK056203.1 - 21 1913
AK056305.1 - 10 862
AK091589.1 - 21 2019
AK097094.1 - 2 1472
AK226106.1 - 22 1998
AY358830.1 - 22 1977
BC009239.2 - 21 1717 2988664

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

protein polyubiquitination
autophagy
protein catabolic process
negative regulation of endocytosis
viral budding
protein autoubiquitination
ubiquitin-dependent endocytosis
positive regulation of xenophagy
positive regulation of autophagosome assembly
ubiquitin-protein transferase activity
protein binding
zinc ion binding
ligase activity
ubiquitin protein ligase activity

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-blue-67f85b6989-dz54l:80/100.66.79.164:80.
git-commit: a6f50562c839d11f1e5940093fdcbf4cc66353c1
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.1-Offline