This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other CCDC74A SIRNA Assays ›
Gene Symbol:
CCDC74A CCDC74B
Gene Name:
coiled-coil domain containing 74A
coiled-coil domain containing 74B
Gene Aliases:
-
-
Chromosome Location:
Chr.2: 131522621 - 131533666 on Build GRCh38
Chromosome Location:
Chr.2: 130139283 - 130145134 on Build GRCh38
UniGene ID:
Hs.351461
Hs.29383
Species:
Human
Species Specific ID (Flybase ID):
-
-
siRNA ID 36946

Purification
Size
Availability Inventoried
Catalog # AM16708
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: CCDC74A
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001258306.1 NP_001245235.1 4, 5 625
NM_138770.2 NP_620125.1 4, 5 823
XM_005263835.3 XP_005263892.1 4, 5 723
XM_005263836.4 XP_005263893.1 2, 3 1106
XM_005263837.1 XP_005263894.1 4, 5 741
XM_006712828.3 XP_006712891.1 4, 5 922
XM_017005234.1 XP_016860723.1 4, 5 935
XM_017005235.1 XP_016860724.1 4, 5 994
XM_017005236.1 XP_016860725.1 4, 5 736
XM_017005239.1 XP_016860728.1 4, 5 796
XM_017005241.1 XP_016860730.1 5, 6 731
XM_017005244.1 XP_016860733.1 5, 6 527
XM_017005245.1 XP_016860734.1 4, 5 753
GenBank mRNA BC016861.1 - 4, 5 738 3914851
BC063387.1 - 4, 5 562 5494806
Gene Symbol: CCDC74B
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001258307.1 NP_001245236.1 4, 5 626
NM_207310.2 NP_997193.1 4, 5 824
XM_005263840.2 XP_005263897.1 4, 5 849
XM_005263842.2 XP_005263899.1 4, 5 651
XM_006712833.2 XP_006712896.1 4, 5 861
XM_006712834.3 XP_006712897.1 4, 5 801
XM_006712835.2 XP_006712898.1 4, 5 663
XM_006712837.2 XP_006712900.1 4, 5 603
XM_011512142.2 XP_011510444.1 2, 3 1092
GenBank mRNA AK057609.1 - 4, 5 748
AK297246.1 - 3, 4 1736
BC067771.1 - 4, 5 571 4824929

Back To Top

More Information


Additional Information:

This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-blue-67f85b6989-b4dkc:80/100.66.78.154:80.
git-commit: a6f50562c839d11f1e5940093fdcbf4cc66353c1
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.1-Offline