This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other VWA3B SIRNA Assays ›
Gene Symbol:
VWA3B
Gene Name:
von Willebrand factor A domain containing 3B
Gene Aliases:
SCAR22
Chromosome Location:
Chr.2: 98087132 - 98330675 on Build GRCh38
UniGene ID:
Hs.269977
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID 37684

Purification
Size
Availability Inventoried
Catalog # AM16708
Price 4,126.00
Your Price
Online offer:
4126.0
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: VWA3B
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_144992.4 NP_659429.4 14 2213
XM_005263897.2 XP_005263954.1 14 2178
XM_006712357.2 XP_006712420.1 13 1927
XM_006712359.3 XP_006712422.1 5 607
XM_006712360.3 XP_006712423.1 2 254
XM_011510770.1 XP_011509072.1 14 2178
XM_011510771.2 XP_011509073.1 13 2052
XM_011510772.1 XP_011509074.1 14 2178
XM_011510774.1 XP_011509076.1 14 2178
XM_017003560.1 XP_016859049.1 8 1044
XM_017003561.1 XP_016859050.1 8 1050
XM_017003562.1 XP_016859051.1 8 1100
XM_017003563.1 XP_016859052.1 8 1050
XM_017003564.1 XP_016859053.1 14 2178
GenBank mRNA AK057248.1 - 11 1714
AK090786.1 - 2 180
AK093084.1 - 2 180
AK295743.1 - 8 1108
AL832635.2 - 15 2156
AL832761.1 - 14 2234
AL834173.1 - 5 612
AL834385.1 - 4 476
BC026091.2 - 14 2028
BC095480.1 - 15 2389
BC150653.1 - 14 2153 9057024

Back To Top

More Information



Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-blue-55586bc97b-pwq5j:80/100.66.76.31:80.
git-commit: d8c775737291b1e1a3c617895b4ca460ad723db2
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.1-2025.09.37-1.0