This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other C11ORF45 SIRNA Assays ›
Gene Symbol:
C11orf45
Gene Name:
chromosome 11 open reading frame 45
Gene Aliases:
-
Chromosome Location:
Chr.11: 128899565 - 128909520 on Build GRCh38
UniGene ID:
Hs.689860
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID 37784

Purification
Size
Availability Inventoried
Catalog # AM16708
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: C11orf45
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001256088.1 NP_001243017.1 3 414
NM_145013.2 NP_659450.1 3 403
NR_045767.1 3 337
XM_011542654.2 XP_011540956.1 3 333
XM_011542655.2 XP_011540957.1 3 295
GenBank mRNA AK125634.1 - 3 311
AK314137.1 - 3 380
BC025756.1 - 3 403 5201867

Back To Top

More Information



Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-f5rhb:80/100.66.76.55:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0