This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other AURKC SIRNA Assays ›
Gene Symbol:
AURKC
Gene Name:
aurora kinase C
Gene Aliases:
AIE2, AIK3, ARK3, AurC, HEL-S-90, SPGF5, STK13, aurora-C
Chromosome Location:
Chr.19: 57230317 - 57235550 on Build GRCh38
UniGene ID:
Hs.98338
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID 377

Purification
Size
Availability Inventoried
Catalog # AM16708
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: AURKC
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001015878.1 NP_001015878.1 5, 6 784
NM_001015879.1 NP_001015879.1 5, 6 643
NM_003160.2 NP_003151.2 5, 6 648
GenBank mRNA AB017332.1 - 5, 6 789
AF054621.1 - 5, 6 789
AF059681.1 - 5, 6 844
AK301886.1 - 4, 5 1229
AY661554.1 - 5, 6 643
AY714054.1 - 5, 6 586
BC075064.2 - 4, 5 537 30915373
EU668345.1 - 5, 6 844

Back To Top

More Information


Additional Information:

This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.

Panther Classification:

Molecular Function -

non-receptor serine/threonine protein kinase

Gene Ontology Categories:

Function(s) Process(es)

cytokinesis
protein phosphorylation
spermatogenesis
attachment of spindle microtubules to kinetochore
histone modification
positive regulation of cytokinesis
histone-serine phosphorylation
oocyte development
mitotic spindle midzone assembly
meiotic cell cycle
protein kinase activity
protein serine/threonine/tyrosine kinase activity
ATP binding
histone serine kinase activity

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-blue-67f85b6989-ztldj:80/100.66.73.158:80.
git-commit: a6f50562c839d11f1e5940093fdcbf4cc66353c1
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.1-Offline