siRNA Details
| Gene Symbol: DNAJB7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_145174.1 | NP_660157.1 | 1 | 302 | ||
| GenBank mRNA | AF085232.1 | - | 1 | 302 | ||
| BC112135.1 | - | 1 | 261 | 8327603 | ||
| CR456468.1 | - | 1 | 207 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

DNAJB7| Gene Symbol: DNAJB7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_145174.1 | NP_660157.1 | 1 | 302 | ||
| GenBank mRNA | AF085232.1 | - | 1 | 302 | ||
| BC112135.1 | - | 1 | 261 | 8327603 | ||
| CR456468.1 | - | 1 | 207 | |||