This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other ZNF785 SIRNA Assays ›
Gene Symbol:
ZNF785
Gene Name:
zinc finger protein 785
Gene Aliases:
-
Chromosome Location:
Chr.16: 30573219 - 30586038 on Build GRCh38
UniGene ID:
Hs.513509
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID 38877

Purification
Size
Availability Inventoried
Catalog # AM16708
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: ZNF785
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_152458.6 NP_689671.2 3 509
XM_011545752.2 XP_011544054.1 3 815
XM_011545753.2 XP_011544055.1 3 815
XM_017022967.1 XP_016878456.1 3 776
GenBank mRNA AK056692.1 - 3 465
AK292465.1 - 3 393
AK298848.1 - 3 488
AK309511.1 - 3 266
BC040642.2 - 3 427 4811661
CN401082.1 - 3 509

Back To Top

More Information


Panther Classification:

Molecular Function -

C2H2 zinc finger transcription factor

Gene Ontology Categories:

Function(s) Process(es)

transcription, DNA-templated
regulation of transcription, DNA-templated
DNA binding
protein binding
metal ion binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-blue-67f85b6989-fh6lz:80/100.66.76.113:80.
git-commit: a6f50562c839d11f1e5940093fdcbf4cc66353c1
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.1-Offline