siRNA Details
| Gene Symbol: LINC00905 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_024335.2 | 5 | 1085 | |||
| NR_110321.1 | 5 | 795 | ||||
| GenBank mRNA | AK058057.1 | - | 5 | 1044 | ||
| BC031284.1 | - | 5 | 795 | |||
| BC069223.1 | - | 5 | 1017 | |||
| BC104221.2 | - | 4 | 982 | |||

LINC00905
Genomic Map