siRNA Details
| Gene Symbol: SNRNP48 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_152551.3 | NP_689764.3 | 1, 2 | 223 | ||
| XM_011514312.2 | XP_011512614.1 | 1, 2 | 159 | |||
| GenBank mRNA | AK056796.1 | - | 1, 2 | 200 | ||
| AK292409.1 | - | 1, 2 | 201 | |||
| AL832462.1 | - | 1, 2 | 224 | |||
| BC054008.1 | - | 1, 2 | 196 | |||
| BC062458.1 | - | 1, 2 | 185 | |||
| BC114630.1 | - | 1, 2 | 191 | 40084896 | ||
| BC114631.1 | - | 1, 2 | 192 | 40084903 | ||

SNRNP48
Genomic Map