This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other TTC39B SIRNA Assays ›
Gene Symbol:
TTC39B
Gene Name:
tetratricopeptide repeat domain 39B
Gene Aliases:
C9orf52
Chromosome Location:
Chr.9: 15167204 - 15307360 on Build GRCh38
UniGene ID:
Hs.563630
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID 39163

Purification
Size
Availability Inventoried
Catalog # AM16708
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: TTC39B
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001168339.1 NP_001161811.1 4 432
NM_001168340.1 NP_001161812.1 4 432
NM_001168342.1 NP_001161814.1 2 307
NM_152574.2 NP_689787.2 4 432
XM_011517732.2 XP_011516034.1 3 229
XM_017014310.1 XP_016869799.1 6 795
XM_017014311.1 XP_016869800.1 5 727
XM_017014312.1 XP_016869801.1 3 297
GenBank mRNA AK091187.1 - 4 318
AK093459.1 - 4 499
AK298998.1 - 4 404
AK301185.1 - 4 283
AK301427.1 - 2 223
AK310271.1 - 5 290
AK310482.1 - 4 432
BC042417.1 - 3 299
BC142985.1 - 4 411 8860428
DT217817.1 - 3 404

Back To Top

More Information



Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-blue-55586bc97b-stzfq:80/100.66.77.128:80.
git-commit: d8c775737291b1e1a3c617895b4ca460ad723db2
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.1-2025.09.37-1.0