This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other ZNF92 SIRNA Assays ›
Gene Symbol:
ZNF92
Gene Name:
zinc finger protein 92
Gene Aliases:
HEL-203, HPF12, HTF12, TF12
Chromosome Location:
Chr.7: 65373799 - 65401135 on Build GRCh38
UniGene ID:
Hs.9521
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID 39210

Purification
Size
Availability Inventoried
Catalog # AM16708
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: ZNF92
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001287532.1 NP_001274461.1 3 987
NM_001287533.1 NP_001274462.1 5 1205
NM_001287534.1 NP_001274463.1 2 860
NM_007139.3 NP_009070.2 3 956
NM_152626.3 NP_689839.1 4 1083
GenBank mRNA AK091618.1 - 4 1001
BC036439.1 - 3 879 4817733
BC040594.1 - 4 1025 5269950
BX647875.1 - 2 805
EU668348.1 - 4 1027

Back To Top

More Information


Panther Classification:

Molecular Function -

C2H2 zinc finger transcription factor

Gene Ontology Categories:

Function(s) Process(es)

transcription, DNA-templated
regulation of transcription, DNA-templated
RNA polymerase II core promoter proximal region sequence-specific DNA binding
transcription factor activity, sequence-specific DNA binding
zinc ion binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-blue-67f85b6989-nzkhn:80/100.66.78.154:80.
git-commit: a6f50562c839d11f1e5940093fdcbf4cc66353c1
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.1-Offline