This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other MS4A6A SIRNA Assays ›
Gene Symbol:
MS4A6A
Gene Name:
membrane spanning 4-domains A6A
Gene Aliases:
4SPAN3, 4SPAN3.2, CD20L3, CDA01, MS4A6, MST090, MSTP090
Chromosome Location:
Chr.11: 60171607 - 60184666 on Build GRCh38
UniGene ID:
Hs.523702
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID 39720

Purification
Size
Availability Inventoried
Catalog # AM16708
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: MS4A6A
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001247999.1 NP_001234928.1 5, 6 888
NM_022349.3 NP_071744.2 4, 5 699
NM_152851.2 NP_690590.1 4, 5 699
NM_152852.2 NP_690591.1 5, 6 824
XM_005274177.2 XP_005274234.1 5, 6 883
XM_006718658.2 XP_006718721.1 5, 6 758
XM_006718660.2 XP_006718723.1 4, 5 623
XM_006718661.2 XP_006718724.1 4, 5 623
XM_017018123.1 XP_016873612.1 3, 4 359
XM_017018124.1 XP_016873613.1 3, 4 359
GenBank mRNA AB013104.1 - 4, 5 407
AF212240.1 - 4, 5 568
AF237908.1 - 4, 5 569
AF253977.1 - 4, 5 569
AF261136.1 - 4, 5 569
AF286866.1 - 3, 4 347
AF354930.1 - 4, 5 569
AK291870.1 - 5, 6 824
AK301387.1 - 3, 4 259
BC108728.1 - 4, 5 472 6196213
DA917243.1 - 4, 5 523
DA979138.1 - 4, 5 470

Back To Top

More Information


Panther Classification:

Molecular Function -

transporter

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-blue-67f85b6989-7p2bx:80/100.66.78.154:80.
git-commit: a6f50562c839d11f1e5940093fdcbf4cc66353c1
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.1-Offline