siRNA Details
| Gene Symbol: ZNF548 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | BC030788.2 | - | 3 | 426 | 5266647 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

ZNF548| Gene Symbol: ZNF548 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | BC030788.2 | - | 3 | 426 | 5266647 | |