siRNA Details
| Gene Symbol: PROSER2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_153256.3 | NP_694988.3 | 4 | 1542 | ||
| XM_011519438.2 | XP_011517740.1 | 5 | 1861 | |||
| XM_017016028.1 | XP_016871517.1 | 4 | 2528 | |||
| GenBank mRNA | BC007547.1 | - | 1 | 460 | ||
| BC017269.2 | - | 5 | 1260 | 4588686 | ||
| BC029134.1 | - | 1 | 843 | |||
| BC048800.1 | - | 4 | 1542 | 5215506 | ||

PROSER2
Genomic Map