siRNA Details
| Gene Symbol: TMEM86A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_153347.2 | NP_699178.1 | 2 | 424 | ||
| GenBank mRNA | AK074600.1 | - | 2 | 380 | ||
| BC017073.2 | - | 1 | 212 | |||
| BC035692.1 | - | 2 | 355 | 5759114 | ||
| DA047874.1 | - | 2 | 424 | |||

TMEM86A
Genomic Map