This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other ANKFN1 SIRNA Assays ›
Gene Symbol:
ANKFN1
Gene Name:
ankyrin repeat and fibronectin type III domain containing 1
Gene Aliases:
-
Chromosome Location:
Chr.17: 56110935 - 56517016 on Build GRCh38
UniGene ID:
Hs.673040
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID 40086

Purification
Size
Availability Inventoried
Catalog # AM16708
Price 4,126.00
Your Price
Online offer:
4126.0
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: ANKFN1
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_153228.2 NP_694960.2 3 150
XM_006721728.3 XP_006721791.1 2 373
XM_011524428.2 XP_011522730.1 3 150
XM_011524429.2 XP_011522731.1 3 307
XM_011524430.2 XP_011522732.1 4 273
XM_011524431.2 XP_011522733.1 3 205
XM_017024263.1 XP_016879752.1 5 367
XM_017024264.1 XP_016879753.1 5 1352
XM_017024265.1 XP_016879754.1 5 561
XM_017024266.1 XP_016879755.1 3 322
XM_017024267.1 XP_016879756.1 2 174
XM_017024268.1 XP_016879757.1 4 1258
XM_017024269.1 XP_016879758.1 2 447
XM_017024270.1 XP_016879759.1 5 365
GenBank mRNA AK095654.1 - 3 150

Back To Top

More Information



Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-zpsr4:80/100.66.78.116:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0