This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other BEST4 SIRNA Assays ›
Gene Symbol:
BEST4
Gene Name:
bestrophin 4
Gene Aliases:
VMD2L2
Chromosome Location:
Chr.1: 44781836 - 44792828 on Build GRCh38
UniGene ID:
Hs.302513
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID 40209

Purification
Size
Availability Inventoried
Catalog # AM16708
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: BEST4
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_153274.2 NP_695006.1 9 1943
XM_017001022.1 XP_016856511.1 11 3294
GenBank mRNA AF440757.1 - 9 1894
BM142321.1 - 1 357

Back To Top

More Information


Panther Classification:

Molecular Function -

ion channel

Gene Ontology Categories:

Function(s) Process(es)

biological_process
chloride transmembrane transport
molecular_function
chloride channel activity

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-blue-67f85b6989-47mq8:80/100.66.76.147:80.
git-commit: a6f50562c839d11f1e5940093fdcbf4cc66353c1
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.1-Offline