siRNA Details
| Gene Symbol: FNDC5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001171940.1 | NP_001165411.2 | 4 | 489 | ||
| NM_001171941.2 | NP_001165412.1 | 4 | 423 | |||
| NM_153756.2 | NP_715637.2 | 4 | 489 | |||
| GenBank mRNA | AK092102.1 | - | 4 | 422 | ||
| BC062297.1 | - | 4 | 412 | 6338770 | ||
| BE502835.1 | - | 4 | 490 | |||
| BX537781.1 | - | 4 | 365 | |||
| DA393478.1 | - | 2 | 93 | |||

FNDC5
Genomic Map