This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other DOK7 SIRNA Assays ›
Gene Symbol:
DOK7
Gene Name:
docking protein 7
Gene Aliases:
C4orf25, CMS10, CMS1B
Chromosome Location:
Chr.4: 3463306 - 3501476 on Build GRCh38
UniGene ID:
Hs.122110
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID 41123

Purification
Size
Availability Inventoried
Catalog # AM16708
Price 4,126.00
Your Price
Online offer:
4126.0
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: DOK7
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001164673.1 NP_001158145.1 7 2120
NM_001256896.1 NP_001243825.1 4 1829
NM_173660.4 NP_775931.3 7 2131
GenBank mRNA AK091037.1 - 7 2115
AK096456.1 - 4 1829
BC043568.1 - 3 1467
BC062369.1 - 3 1523
BC131544.1 - 7 2067 40085498
BC141852.1 - 7 2102 8860352

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

positive regulation of protein tyrosine kinase activity
insulin receptor binding
lipid binding
protein kinase binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-l4hk6:80/100.66.76.55:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0