siRNA Details
| Gene Symbol: RASGEF1C | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_175062.3 | NP_778232.2 | 6 | 783 | ||
| GenBank mRNA | AF449764.1 | - | 5 | 971 | ||
| AK093160.1 | - | 3 | 635 | |||
| BC036802.1 | - | 7 | 801 | |||
| BC057759.1 | - | 3 | 437 | 5268021 | ||

RASGEF1C
Genomic Map