siRNA Details
| Gene Symbol: HIST1H2BF | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_003522.3 | NP_003513.1 | 1 | 379 | ||
| GenBank mRNA | BC096120.1 | - | 1 | 379 | 40002352 | |
| BC096123.1 | - | 1 | 379 | 40002357 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

HIST1H2BF| Gene Symbol: HIST1H2BF | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_003522.3 | NP_003513.1 | 1 | 379 | ||
| GenBank mRNA | BC096120.1 | - | 1 | 379 | 40002352 | |
| BC096123.1 | - | 1 | 379 | 40002357 | ||