siRNA Details
| Gene Symbol: HIST1H4E | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_003545.3 | NP_003536.1 | 1 | 248 | ||
| GenBank mRNA | AK312153.1 | - | 1 | 341 | ||
| BC012587.1 | - | 1 | 287 | |||
| BC050615.1 | - | 1 | 264 | |||
| CR542180.1 | - | 1 | 248 | |||
| CR542189.1 | - | 1 | 248 | |||

HIST1H4E
Genomic Map