This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other LCE2B SIRNA Assays ›
Gene Symbol:
LCE2B
Gene Name:
late cornified envelope 2B
Gene Aliases:
LEP10, SPRL1B, XP5
Chromosome Location:
Chr.1: 152686123 - 152687401 on Build GRCh38
UniGene ID:
Hs.234766
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID 42312

Purification
Size
Availability Inventoried
Catalog # AM16708
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: LCE2B
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_014357.4 NP_055172.1 2 409
GenBank mRNA AF086289.1 - 2 409
BC117235.1 - 2 400
BC117237.1 - 2 400

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

epidermis development
peptide cross-linking
keratinocyte differentiation
keratinization
molecular_function
structural molecule activity

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-blue-67f85b6989-nzkhn:80/100.66.78.154:80.
git-commit: a6f50562c839d11f1e5940093fdcbf4cc66353c1
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.1-Offline