This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other DUSP23 SIRNA Assays ›
Gene Symbol:
DUSP23
Gene Name:
dual specificity phosphatase 23
Gene Aliases:
DUSP25, LDP-3, MOSP, VHZ
Chromosome Location:
Chr.1: 159780946 - 159782546 on Build GRCh38
UniGene ID:
Hs.425801
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID 42513

Purification
Size
Availability Inventoried
Catalog # AM16708
Price 4,126.00
Your Price
Online offer:
4126.0
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: DUSP23
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001319658.1 NP_001306587.1 2 629
NM_001319659.1 NP_001306588.1 3 588
NM_017823.4 NP_060293.2 3 581
GenBank mRNA AB164404.1 - 2 614
AF086553.1 - 2 429
AK000449.1 - 3 565
BC001140.1 - 3 551 3535406
BG823348.1 - 2 611
BQ679163.1 - 3 600

Back To Top

More Information


Panther Classification:

Molecular Function -

protein phosphatase

Gene Ontology Categories:

Function(s) Process(es)

dephosphorylation
peptidyl-tyrosine dephosphorylation
protein serine/threonine phosphatase activity
protein tyrosine phosphatase activity
protein binding
protein tyrosine/serine/threonine phosphatase activity
phosphatase activity

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-blue-98cc9b549-mbm4f:80/100.66.76.81:80.
git-commit: 5e7f4b920385fdbbb5f8d978f1c7c2eb3f871195
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.40.1-Offline