This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other DSCR8 SIRNA Assays ›
Gene Symbol:
DSCR8
Gene Name:
Down syndrome critical region 8
Gene Aliases:
C21orf65, CT25.1a, CT25.1b, MMA-1, MMA-1a, MMA-1b, MMA1, MTAG2
Chromosome Location:
Chr.21: 38121451 - 38156511 on Build GRCh38
UniGene ID:
Hs.733345
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID 42608

Purification
Size
Availability Inventoried
Catalog # AM16708
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: DSCR8
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NR_026838.1 3 528
NR_026840.1 3 289
NR_026841.1 3 271
GenBank mRNA AF321193.1 - 3 246
AF426256.1 - 3 194
AJ001904.1 - 3 231
AJ297914.2 - 3 249
AJ306839.2 - 3 267
AJ306840.2 - 3 321
AJ783421.1 - 3 506
BC117304.1 - 3 203
BC117330.1 - 3 203
BC143995.1 - 3 221

Back To Top

More Information


Additional Information:

This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.

Gene Ontology Categories:

Function(s) Process(es)

biological_process
molecular_function
protein binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-blue-67f85b6989-kbrl8:80/100.66.76.113:80.
git-commit: a6f50562c839d11f1e5940093fdcbf4cc66353c1
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.1-Offline