This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other KRTAP4-5 SIRNA Assays ›
Gene Symbol:
KRTAP4-5
Gene Name:
keratin associated protein 4-5
Gene Aliases:
KAP4.5, KRTAP4.5
Chromosome Location:
Chr.17: 41148924 - 41149802 on Build GRCh38
UniGene ID:
Hs.514863
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID 42618

Purification
Size
Availability Inventoried
Catalog # AM16708
Price 4,126.00
Your Price
Online offer:
4126.0
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: KRTAP4-5
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_033188.3 NP_149445.3 1 656
GenBank mRNA AJ406937.1 - 1 671

Back To Top

More Information


Gene Ontology Categories:

Function(s)

aging
hair cycle

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-green-c565d5cb7-m6gg6:80/100.66.76.251:80.
git-commit: 04d74f786bdbf98a9bc1b2a22061fbbfd0e8b2c2
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.38.0-2025.10.43-1.0