This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other GLI4 SIRNA Assays ›
Gene Symbol:
GLI4 ZFP41
Gene Name:
GLI family zinc finger 4
ZFP41 zinc finger protein
Gene Aliases:
HKR4, ZNF928
ZNF753, zfp-41
Chromosome Location:
Chr.8: 143267437 - 143276931 on Build GRCh38
Chromosome Location:
Chr.8: 143246821 - 143262705 on Build GRCh38
UniGene ID:
Hs.400533
Hs.668016
Species:
Human
Species Specific ID (Flybase ID):
-
-
siRNA ID 42805

Purification
Size
Availability Inventoried
Catalog # AM16708
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: GLI4
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_138465.3 NP_612474.1 4 663
GenBank mRNA AY129020.1 - 1 1653
BC014165.2 - 3 629 4549240
Gene Symbol: ZFP41
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
GenBank mRNA AL831951.1 - 4 2745

Back To Top

More Information


Panther Classification:

Molecular Function -

C2H2 zinc finger transcription factor

Gene Ontology Categories:

Function(s) Process(es)

regulation of transcription, DNA-templated
biological_process
molecular_function
DNA binding
transcription factor activity, sequence-specific DNA binding
metal ion binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-blue-67f85b6989-gmb9z:80/100.66.79.164:80.
git-commit: a6f50562c839d11f1e5940093fdcbf4cc66353c1
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.1-Offline