This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other H2AFJ SIRNA Assays ›
Gene Symbol:
H2AFJ
Gene Name:
H2A histone family member J
Gene Aliases:
H2AJ
Chromosome Location:
Chr.12: 14774336 - 14778002 on Build GRCh38
UniGene ID:
Hs.524280
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID 42994

Purification
Size
Availability Inventoried
Catalog # AM16708
Price 4,126.00
Your Price
Online offer:
4126.0
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: H2AFJ
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_177925.3 NP_808760.1 1 419
NR_027716.2 1 419
GenBank mRNA AK001765.1 - 1 350
AK303301.1 - 2 296
AL133626.1 - 1 173
BC003602.1 - 1 340 3354610
BM689354.1 - 1 423
BU594879.1 - 1 397
DA808847.1 - 1 420
DB084616.1 - 1 420

Back To Top

More Information


Panther Classification:

Molecular Function -

histone

Gene Ontology Categories:

Function(s) Process(es)

chromatin silencing
biological_process
molecular_function
DNA binding
protein heterodimerization activity

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-blue-67f85b6989-xg9l8:80/100.66.77.189:80.
git-commit: a6f50562c839d11f1e5940093fdcbf4cc66353c1
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.1-Offline