This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other CCDC116 SIRNA Assays ›
Gene Symbol:
CCDC116
Gene Name:
coiled-coil domain containing 116
Gene Aliases:
-
Chromosome Location:
Chr.22: 21632121 - 21637327 on Build GRCh38
UniGene ID:
Hs.131615
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID 43885

Purification
Size
Availability Inventoried
Catalog # AM16708
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: CCDC116
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_152612.2 NP_689825.2 3 645
XM_006724159.2 XP_006724222.1 3 864
XM_011529984.2 XP_011528286.1 2 1187
XM_011529985.1 XP_011528287.1 3 864
GenBank mRNA AK093365.1 - 3 713
AK302030.1 - 3 726
BC033499.1 - 3 645 5172053

Back To Top

More Information


Gene Ontology Categories:

Process(es)

protein binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-green-55658555d6-qfdf2:80/100.66.76.55:80.
git-commit: ec8e7df5f8fec8d765bd419205f1b5046a016d37
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.41.0-Offline