This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other MAP10 SIRNA Assays ›
Gene Symbol:
MAP10
Gene Name:
microtubule associated protein 10
Gene Aliases:
KIAA1383, MTR120
Chromosome Location:
Chr.1: 232804892 - 232810346 on Build GRCh38
UniGene ID:
Hs.160373
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID 44043

Purification
Size
Availability Inventoried
Catalog # AM16708
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: MAP10
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_019090.2 NP_061963.2 1 1298
GenBank mRNA AB037804.1 - 1 747
AK290216.1 - 1 824
BC036663.2 - 1 776 5266476
BC109122.2 - 1 1164 40036000

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

cytoplasmic microtubule organization
positive regulation of cytokinesis
regulation of microtubule-based process
mitotic spindle midzone assembly
cell division
microtubule binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-blue-67f85b6989-f8k5r:80/100.66.76.16:80.
git-commit: a6f50562c839d11f1e5940093fdcbf4cc66353c1
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.1-Offline