This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other NLRX1 SIRNA Assays ›
Gene Symbol:
NLRX1
Gene Name:
NLR family member X1
Gene Aliases:
CLR11.3, DLNB26, NOD26, NOD5, NOD9
Chromosome Location:
Chr.11: 119168334 - 119184016 on Build GRCh38
UniGene ID:
Hs.524082
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID 44469

Purification
Size
Availability Inventoried
Catalog # AM16708
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: NLRX1
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001282143.1 NP_001269072.1 7 1996
NM_001282144.1 NP_001269073.1 7 2404
NM_001282358.1 NP_001269287.1 7 1928
NM_024618.3 NP_078894.2 7 2023
XM_005271669.1 XP_005271726.1 7 2247
XM_005271670.1 XP_005271727.1 6 1900
XM_005271671.1 XP_005271728.1 6 2074
XM_005271672.1 XP_005271729.1 6 1897
XM_006718904.1 XP_006718967.1 7 2281
XM_011542980.1 XP_011541282.1 7 1911
XM_017018294.1 XP_016873783.1 8 2342
GenBank mRNA AB094095.1 - 7 2007
AK025131.1 - 2 650
AK056454.1 - 6 1721
AK095247.1 - 5 1208
AK225223.1 - 2 650
AK291716.1 - 7 1990
AK303002.1 - 6 1897
AY245437.1 - 6 1766
BC013199.1 - 3 1127
BC110890.1 - 7 2369 4937963
BX647705.1 - 7 1981
EF452237.1 - 6 1766

Back To Top

More Information


Panther Classification:

Molecular Function -

scaffold/adaptor protein

Gene Ontology Categories:

Function(s) Process(es)

viral process
negative regulation of type I interferon production
negative regulation of interferon-beta production
negative regulation of interleukin-6 production
negative regulation of RIG-I signaling pathway
negative regulation of I-kappaB kinase/NF-kappaB signaling
innate immune response
negative regulation of innate immune response
negative regulation of inflammatory response
protein binding
ATP binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-blue-67f85b6989-f8k5r:80/100.66.76.16:80.
git-commit: a6f50562c839d11f1e5940093fdcbf4cc66353c1
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.1-Offline