This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other PFKM SIRNA Assays ›
Gene Symbol:
PFKM
Gene Name:
phosphofructokinase, muscle
Gene Aliases:
ATP-PFK, GSD7, PFK-1, PFK1, PFKA, PFKX, PPP1R122
Chromosome Location:
Chr.12: 48105278 - 48146404 on Build GRCh38
UniGene ID:
Hs.75160
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID 44

Purification
Size
Availability Inventoried
Catalog # AM16708
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: PFKM
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_000289.5 NP_000280.1 2, 3 278
NM_001166686.1 NP_001160158.1 4, 5 580
NM_001166687.1 NP_001160159.1 2, 3 132
NM_001166688.1 NP_001160160.1 1, 2 221
XM_005268974.1 XP_005269031.1 5, 6 477
XM_005268975.1 XP_005269032.1 5, 6 616
XM_005268976.3 XP_005269033.1 5, 6 659
XM_005268977.1 XP_005269034.1 4, 5 463
XM_005268978.2 XP_005269035.1 4, 5 376
XM_005268979.1 XP_005269036.1 4, 5 463
XM_011538487.1 XP_011536789.1 4, 5 411
XM_011538488.2 XP_011536790.1 2, 3 243
XM_017019467.1 XP_016874956.1 5, 6 546
XM_017019468.1 XP_016874957.1 4, 5 398
XM_017019469.1 XP_016874958.1 4, 5 463
GenBank mRNA AK126229.1 - 5, 6 530
AK312284.1 - 2, 3 160
BC000534.2 - 1, 2 108 2964710
BC007798.2 - 1, 2 88
BC012799.2 - 1, 2 108 2964710
BC013298.2 - 1, 2 108 2964710
BC021203.2 - 2, 3 130 4100117
DA525437.1 - 2, 3 278
DA560355.1 - 2, 3 132
DA761651.1 - 1, 2 221
DA795106.1 - 2, 3 263
KU178251.1 - 1, 2 93
KU178252.1 - 1, 2 93
KU178253.1 - 1, 2 93
M26066.1 - 2, 3 169
U24183.1 - 2, 3 169
Y00698.1 - 1, 2 112

Back To Top

More Information


Additional Information:

This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.

Panther Classification:

Molecular Function -

carbohydrate kinase kinase transferase metabolite interconversion enzyme

Gene Ontology Categories:

Function(s) Process(es)

glycogen catabolic process
fructose 6-phosphate metabolic process
glycolytic process
positive regulation of insulin secretion
glucose homeostasis
muscle cell cellular homeostasis
carbohydrate phosphorylation
protein oligomerization
canonical glycolysis
glycolysis from storage polysaccharide through glucose-1-phosphate
6-phosphofructokinase activity
protein binding
ATP binding
protein C-terminus binding
kinase binding
identical protein binding
protein homodimerization activity
metal ion binding
fructose binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-blue-67f85b6989-dz54l:80/100.66.79.164:80.
git-commit: a6f50562c839d11f1e5940093fdcbf4cc66353c1
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.1-Offline