siRNA Details
| Gene Symbol: ZNFX1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_021035.2 | NP_066363.1 | 14 | 3714 | ||
| XM_006723836.1 | XP_006723899.1 | 14 | 3544 | |||
| XM_011528924.1 | XP_011527226.1 | 13 | 3589 | |||
| XM_017027971.1 | XP_016883460.1 | 12 | 2640 | |||
| GenBank mRNA | AB037825.2 | - | 14 | 3532 | ||
| AK023836.1 | - | 10 | 1466 | |||

ZNFX1
Genomic Map