This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other NECAB3 SIRNA Assays ›
Gene Symbol:
NECAB3
Gene Name:
N-terminal EF-hand calcium binding protein 3
Gene Aliases:
APBA2BP, EFCBP3, NIP1, STIP3, SYTIP2, XB51, dJ63M2.4, dJ63M2.5
Chromosome Location:
Chr.20: 33657087 - 33674458 on Build GRCh38
UniGene ID:
Hs.516986
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID 45602

Purification
Size
Availability Inventoried
Catalog # AM16708
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: NECAB3
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_031231.3 NP_112508.3 10, 11 990
NM_031232.3 NP_112509.3 9, 10 1092
XM_005260510.1 XP_005260567.1 9, 10 906
XM_011528991.1 XP_011527293.1 6, 7 616
XM_017028015.1 XP_016883504.1 6, 7 615
XM_017028016.1 XP_016883505.1 6, 7 544
GenBank mRNA AB039947.1 - 7, 8 794
AF193759.1 - 10, 11 897
AF409141.1 - 10, 11 938
AK023706.1 - 9, 10 1196
AK124388.1 - 6, 7 1064
AK291895.1 - 10, 11 920
BC047673.1 - 10, 11 945 5755231
BQ212085.1 - 5, 6 688

Back To Top

More Information


Additional Information:

This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.

Gene Ontology Categories:

Function(s) Process(es)

protein secretion
protein metabolic process
regulation of amyloid precursor protein biosynthetic process
calcium ion binding
protein binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-blue-67f85b6989-d7bgt:80/100.66.77.211:80.
git-commit: a6f50562c839d11f1e5940093fdcbf4cc66353c1
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.1-Offline