This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other TMEM190 SIRNA Assays ›
Gene Symbol:
TMEM190
Gene Name:
transmembrane protein 190
Gene Aliases:
MDAC1
Chromosome Location:
Chr.19: 55376126 - 55378246 on Build GRCh38
UniGene ID:
Hs.590943
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID 45694

Purification
Size
Availability Inventoried
Catalog # AM16708
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: TMEM190
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_139172.1 NP_631911.1 2, 3 121
GenBank mRNA AF442729.1 - 2, 3 121
BC128189.1 - 2, 3 121 40113868
BC128190.1 - 2, 3 121 40113870

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

hematopoietic progenitor cell differentiation
protein binding
protein self-association

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-blue-57b588f4f5-bgnf4:80/100.66.76.30:80.
git-commit: 377debcfccf3d85e21e16e6539a0b18561fa37ae
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.39.0-Offline