This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other RBMXL3 SIRNA Assays ›
Gene Symbol:
RBMXL3
Gene Name:
RNA binding motif protein, X-linked like 3
Gene Aliases:
CXorf55
Chromosome Location:
Chr.X: 115189400 - 115192868 on Build GRCh38
UniGene ID:
Hs.376008
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID 45733

Purification
Size
Availability Inventoried
Catalog # AM16708
Price 283.00
Your Price
Online offer: 209.42
209.42
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: RBMXL3
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001145346.1 NP_001138818.1 1 1654
GenBank mRNA AK097568.1 - 1 677
AK301904.1 - 1 1654

Back To Top

More Information


Panther Classification:

Molecular Function -

RNA splicing factor

Gene Ontology Categories:

Process(es)

nucleotide binding
RNA binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-green-c565d5cb7-bwr29:80/100.66.78.44:80.
git-commit: 04d74f786bdbf98a9bc1b2a22061fbbfd0e8b2c2
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.38.0-2025.10.43-1.0