This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other SPCS1 SIRNA Assays ›
Gene Symbol:
SPCS1
Gene Name:
signal peptidase complex subunit 1
Gene Aliases:
HSPC033, SPC1, SPC12, YJR010C-A
Chromosome Location:
Chr.3: 52705841 - 52708182 on Build GRCh38
UniGene ID:
Hs.11125
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID 46204

Purification
Size
Availability Inventoried
Catalog # AM16708
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: SPCS1
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_014041.3 NP_054760.3 3 546
GenBank mRNA AF092138.1 - 3 275
AK027426.1 - 3 275
AX775783.1 - 3 275
AX775785.1 - 4 1060
AX775787.1 - 3 249
AX775833.1 - 3 265
BC000884.1 - 3 249 3462307
BI826431.1 - 3 240
BQ423345.1 - 3 282
L38852.1 - 3 265

Back To Top

More Information


Panther Classification:

Molecular Function -

protease

Gene Ontology Categories:

Function(s) Process(es)

signal peptide processing
proteolysis
protein targeting to ER
molecular_function
protein binding
peptidase activity

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-blue-67f85b6989-vzv8k:80/100.66.76.113:80.
git-commit: a6f50562c839d11f1e5940093fdcbf4cc66353c1
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.1-Offline