This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other PI15 SIRNA Assays ›
Gene Symbol:
PI15
Gene Name:
peptidase inhibitor 15
Gene Aliases:
CRISP8, P24TI, P25TI
Chromosome Location:
Chr.8: 74824537 - 74855044 on Build GRCh38
UniGene ID:
Hs.98558
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID 46244

Purification
Size
Availability Inventoried
Catalog # AM16708
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: PI15
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001324403.1 NP_001311332.1 2 339
NM_015886.4 NP_056970.1 2 311
GenBank mRNA BC074931.2 - 1 155 30915489
BC074932.2 - 1 155 30915266
BC126290.1 - 1 160 8992006
BC126292.1 - 1 162 8992008
CR749657.1 - 2 312
D45027.1 - 1 616

Back To Top

More Information


Panther Classification:

Molecular Function -

defense/immunity protein

Gene Ontology Categories:

Function(s) Process(es)

biological_process
negative regulation of peptidase activity
peptidase inhibitor activity

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-blue-67f85b6989-rdsp8:80/100.66.79.164:80.
git-commit: a6f50562c839d11f1e5940093fdcbf4cc66353c1
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.1-Offline