siRNA Details
| Gene Symbol: MIGA2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_032809.2 | NP_116198.2 | 13 | 1477 | ||
| XM_005252282.1 | XP_005252339.1 | 13 | 1496 | |||
| XM_011519117.1 | XP_011517419.1 | 13 | 1517 | |||
| XM_011519118.1 | XP_011517420.1 | 13 | 1426 | |||
| GenBank mRNA | AK027502.1 | - | 5 | 371 | ||
| AK074127.1 | - | 14 | 1593 | |||
| AK075421.1 | - | 11 | 1489 | |||
| AK301241.1 | - | 10 | 1016 | |||
| AK303036.1 | - | 13 | 1517 | |||
| BC009114.1 | - | 13 | 1477 | 4153683 | ||

MIGA2
Genomic Map