This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other LACRT SIRNA Assays ›
Gene Symbol:
LACRT
Gene Name:
lacritin
Gene Aliases:
-
Chromosome Location:
Chr.12: 54630839 - 54634879 on Build GRCh38
UniGene ID:
Hs.307096
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID 46430

Purification
Size
Availability Inventoried
Catalog # AM16708
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: LACRT
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_033277.1 NP_150593.1 2, 3 149
GenBank mRNA AF238867.1 - 2, 3 163
BC062217.1 - 2, 3 169 30327037
BC069317.1 - 2, 3 149 7262178

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

protein acetylation
positive regulation of cell proliferation
epithelial structure maintenance
positive regulation of macroautophagy
calcium-mediated signaling
negative regulation of lipopolysaccharide-mediated signaling pathway
calcineurin-NFAT signaling cascade
protein localization to Golgi apparatus
defense response to bacterium
negative regulation of apoptotic process
positive regulation of epithelial cell proliferation
positive regulation of peptidyl-tyrosine phosphorylation
positive regulation of calcium-mediated signaling
positive regulation of secretion
positive regulation of release of sequestered calcium ion into cytosol
positive regulation of NFAT protein import into nucleus
positive regulation of epithelial cell proliferation involved in wound healing
tear secretion
glycoprotein binding
fibronectin binding
collagen binding
growth factor activity
laminin-1 binding
protein N-terminus binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-blue-67f85b6989-blq6p:80/100.66.74.192:80.
git-commit: a6f50562c839d11f1e5940093fdcbf4cc66353c1
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.1-Offline