siRNA Details
| Gene Symbol: Bmp6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_013107.1 | NP_037239.1 | 2 | 704 | ||
| GenBank mRNA | AY184240.1 | - | 2 | 704 | ||
| U66298.1 | - | 2 | 477 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Bmp6| Gene Symbol: Bmp6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_013107.1 | NP_037239.1 | 2 | 704 | ||
| GenBank mRNA | AY184240.1 | - | 2 | 704 | ||
| U66298.1 | - | 2 | 477 | |||