This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other BLCAP SIRNA Assays ›
Gene Symbol:
Blcap
Gene Name:
bladder cancer associated protein
Gene Aliases:
-
Chromosome Location:
Chr.3: 154040165 - 154042096 on Build Rnor_6.0
UniGene ID:
Rn.127812
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID 46919

Purification
Size
Availability Inventoried
Catalog # AM16708
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Blcap
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_133582.3 NP_598266.1 1 419
GenBank mRNA AB071598.1 - 1 387
BC087661.1 - 1 212 7313852

Back To Top

More Information


Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_016916, BM950751, AK170338, AK018127, BC083339, and others.

Gene Ontology Categories:

Function(s)

apoptotic nuclear changes

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-l4hk6:80/100.66.76.55:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0