siRNA Details
| Gene Symbol: S100a3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053681.1 | NP_446133.1 | 3 | 410 | ||
| GenBank mRNA | AF140231.1 | - | 3 | 410 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

S100a3| Gene Symbol: S100a3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053681.1 | NP_446133.1 | 3 | 410 | ||
| GenBank mRNA | AF140231.1 | - | 3 | 410 | ||