This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other SLC2A4 SIRNA Assays ›
Gene Symbol:
Slc2a4
Gene Name:
solute carrier family 2 (facilitated glucose transporter), member 4
Gene Aliases:
Glut4
Chromosome Location:
Chr.10: 56552983 - 56558561 on Build Rnor_6.0
UniGene ID:
Rn.1314
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID 48242

Purification
Size
Availability Inventoried
Catalog # AM16708
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Slc2a4
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_012751.1 NP_036883.1 3 371
XM_006246596.2 XP_006246658.1 4 344
GenBank mRNA BC085757.1 - 3 416 7192278
D28561.1 - 3 371
FQ216450.1 - 4 310
FQ232657.1 - 3 430
FQ233419.1 - 3 406
J04524.1 - 3 353
KJ696745.1 - 3 256
M25482.1 - 3 336
X14771.1 - 3 384

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

amylopectin biosynthetic process
glucose transport
cellular response to insulin stimulus
hexose transmembrane transport
glucose homeostasis
response to ethanol
brown fat cell differentiation
transmembrane transport
cellular response to hypoxia
cellular response to osmotic stress
glucose transmembrane transporter activity
protein binding
D-glucose transmembrane transporter activity

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-blue-67f85b6989-xql92:80/100.66.76.113:80.
git-commit: a6f50562c839d11f1e5940093fdcbf4cc66353c1
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.1-Offline