This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other AMBP SIRNA Assays ›
Gene Symbol:
Ambp
Gene Name:
alpha-1-microglobulin/bikunin precursor
Gene Aliases:
-
Chromosome Location:
Chr.5: 78975690 - 78986020 on Build Rnor_6.0
UniGene ID:
Rn.18721
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID 48546

Purification
Size
Availability Inventoried
Catalog # AM16708
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Ambp
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_012901.1 NP_037033.1 1 115
XM_008763759.1 XP_008761981.1 1 201
GenBank mRNA BC088166.1 - 1 100 7368246
S87544.1 - 1 115

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

negative regulation of endopeptidase activity
protein-chromophore linkage
protein catabolic process
serine-type endopeptidase inhibitor activity
IgA binding
heme binding
small molecule binding
protein homodimerization activity

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-green-7669c5b596-k8jnn:80/100.66.77.200:80.
git-commit: 17e8f6c92ed407c129a469238c057b56fdf8abbb
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0