This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other CD44 SIRNA Assays ›
Gene Symbol:
Cd44
Gene Name:
CD44 molecule (Indian blood group)
Gene Aliases:
CD44A, METAA, RHAMM
Chromosome Location:
Chr.3: 92695083 - 92783807 on Build Rnor_6.0
UniGene ID:
Rn.1120
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID 48915

Purification
Size
Availability Inventoried
Catalog # AM16708
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Cd44
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_012924.2 NP_037056.2 8 1080
XM_008762059.1 XP_008760281.1 8 1226
GenBank mRNA AF014365.1 - 3 295
AF295361.1 - 1 50
U96138.1 - 8 1080

Back To Top

More Information


Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_009851, L13611, AJ251594, X69724.

Panther Classification:

Molecular Function -

transmembrane signal receptor

Gene Ontology Categories:

Function(s) Process(es)

regulation of cell growth
branching involved in ureteric bud morphogenesis
blood vessel maturation
wound healing involved in inflammatory response
negative regulation of mature B cell apoptotic process
inflammatory response
cell adhesion
positive regulation of gene expression
response to organic cyclic compound
Wnt signaling pathway
cell migration
hyaluronan catabolic process
neuron projection development
positive regulation of neutrophil apoptotic process
positive regulation of peptidyl-serine phosphorylation
response to vitamin A
positive regulation of heterotypic cell-cell adhesion
macrophage fusion
negative regulation of apoptotic process
negative regulation of cysteine-type endopeptidase activity involved in apoptotic process
negative regulation of DNA damage response, signal transduction by p53 class mediator
cellular response to fibroblast growth factor stimulus
positive regulation of peptidyl-tyrosine phosphorylation
cartilage development
branching involved in prostate gland morphogenesis
positive regulation of ERK1 and ERK2 cascade
monocyte aggregation
positive regulation of monocyte aggregation
negative regulation of intrinsic apoptotic signaling pathway in response to DNA damage by p53 class mediator
hyalurononglucosaminidase activity
transmembrane signaling receptor activity
cytokine receptor activity
epidermal growth factor receptor binding
protein binding
hyaluronic acid binding
protein kinase binding
cytokine binding
phosphoprotein binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-blue-67f85b6989-kbrl8:80/100.66.76.113:80.
git-commit: a6f50562c839d11f1e5940093fdcbf4cc66353c1
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.1-Offline